Methyl-6-methylnicotinate 1-oxide ≥97%

Supplier: Matrix Scientific

083379-1G
102839-622EA 1086.15 USD
102839-622
Methyl-6-methylnicotinate 1-oxide ≥97%
Methyl-6-methylnicotinate 1-oxide

Hazard: Irritant

Formula: C₈H₉NO₃
MDL Number: MFCD22126067
CAS Number: 49668-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR