2-Hydroxy-4-(ethoxycarbonyl)benzoic acid ≥97%

083344-1G
102839-512EA 448.26 USD
102839-512
2-Hydroxy-4-(ethoxycarbonyl)benzoic acid ≥97%
2-Hydroxy-4-(ethoxycarbonyl)benzoic acid

Hazard: Irritant

Formula: C₁₀H₁₀O₅
MDL Number: MFCD11100100
CAS Number: 860709-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR