[[3-(Ethoxycarbonyl)benzyl](methyl)amino]acetic acid ≥97%

Supplier: Matrix Scientific

083313-1G
102839-430EA 949.23 USD
102839-430
[[3-(Ethoxycarbonyl)benzyl](methyl)amino]acetic acid ≥97%
[[3-(Ethoxycarbonyl)benzyl](methyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₄
MDL Number: MFCD22566363
CAS Number: 1417567-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR