Methyl-3,5-difluoro-4-(trifluoromethyl)benzoate ≥97%
Supplier: Matrix Scientific
083312-1G
102839-428EA
949.23
USD
102839-428
Methyl-3,5-difluoro-4-(trifluoromethyl)benzoate ≥97%
Methyl-3,5-difluoro-4-(trifluoromethyl)benzoate
Hazard: Irritant
Formula:
C₉H₅F₅O₂ |
MDL Number:
MFCD22566362 CAS Number: 1417567-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...