Methyl-2-{[(4-methoxybenzyl)amino]methyl}benzoate ≥97%

Supplier: Matrix Scientific

083290-1G
102839-384EA 806.15 USD
102839-384
Methyl-2-{[(4-methoxybenzyl)amino]methyl}benzoate ≥97%
Methyl-2-{[(4-methoxybenzyl)amino]methyl}benzoate

Hazard: Irritant

Formula: C₁₇H₁₉NO₃
MW: 285.35 g/mol
MDL Number: MFCD13790489
CAS Number: 1157652-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR