Methyl-3-(4-methylphenyl)oxirane-2-carboxylate ≥97%

Supplier: Matrix Scientific

083272-1G
102839-336EA 723.08 USD
102839-336
Methyl-3-(4-methylphenyl)oxirane-2-carboxylate ≥97%
Methyl-3-(4-methylphenyl)oxirane-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂O₃
MDL Number: MFCD18089232
CAS Number: 99334-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR