Methyl-3-(3-methylphenyl)oxirane-2-carboxylate ≥97%

Supplier: Matrix Scientific

083271-1G
102839-334EA 723.08 USD
102839-334
Methyl-3-(3-methylphenyl)oxirane-2-carboxylate ≥97%
Methyl-3-(3-methylphenyl)oxirane-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂O₃
MDL Number: MFCD18089231
CAS Number: 126119-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR