Methyl-3-(2-methylphenyl)oxirane-2-carboxylate ≥97%
Supplier: Matrix Scientific
083270-1G
102839-332EA
723.08
USD
102839-332
Methyl-3-(2-methylphenyl)oxirane-2-carboxylate ≥97%
Methyl-3-(2-methylphenyl)oxirane-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₂O₃ |
MDL Number:
MFCD18089230 CAS Number: 1287217-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...