Methyl-3-(3-methoxyphenyl)oxirane-2-carboxylate ≥97%

Supplier: Matrix Scientific

083268-1G
102839-328EA 723.08 USD
102839-328
Methyl-3-(3-methoxyphenyl)oxirane-2-carboxylate ≥97%
Methyl-3-(3-methoxyphenyl)oxirane-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂O₄
MDL Number: MFCD18089229
CAS Number: 99334-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR