Methyl-3-(3-fluorophenyl)oxirane-2-carboxylate ≥97%
Supplier: Matrix Scientific
083265-1G
102839-322EA
723.08
USD
102839-322
Methyl-3-(3-fluorophenyl)oxirane-2-carboxylate ≥97%
Methyl-3-(3-fluorophenyl)oxirane-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₉FO₃ |
MDL Number:
MFCD18089226 CAS Number: 126119-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...