Methyl-3-(3-chlorophenyl)oxirane-2-carboxylate ≥97%
Supplier: Matrix Scientific
083261-1G
102839-312EA
723.08
USD
102839-312
Methyl-3-(3-chlorophenyl)oxirane-2-carboxylate ≥97%
Methyl-3-(3-chlorophenyl)oxirane-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₉ClO₃ |
MDL Number:
MFCD18089223 CAS Number: 133991-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...