Methyl-3-(2-chlorophenyl)oxirane-2-carboxylate ≥97%

Supplier: Matrix Scientific

083260-1G
102839-310EA 723.08 USD
102839-310
Methyl-3-(2-chlorophenyl)oxirane-2-carboxylate ≥97%
Methyl-3-(2-chlorophenyl)oxirane-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₉ClO₃
MDL Number: MFCD18089222
CAS Number: 1258863-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR