Methyl-3-(2-bromophenyl)oxirane-2-carboxylate ≥97%

Supplier: Matrix Scientific

083258-1G
102839-306EA 723.08 USD
102839-306
Methyl-3-(2-bromophenyl)oxirane-2-carboxylate ≥97%
Methyl-3-(2-bromophenyl)oxirane-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₉BrO₃
MDL Number: MFCD18089220
CAS Number: 130089-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR