5,6,7,8-Tetrahydroimidazo[1,2-a]pyridine-3-carbonyl chloride ≥95%

Supplier: Matrix Scientific

083221-1G
102839-202EA 806.15 USD
102839-202
5,6,7,8-Tetrahydroimidazo[1,2-a]pyridine-3-carbonyl chloride ≥95%
5,6,7,8-Tetrahydroimidazo[1,2-a]pyridine-3-carbonyl chloride

Hazard: Irritant

Formula: C₈H₉ClN₂O
MW: 184.63 g/mol
MDL Number: MFCD09258868
CAS Number: 1086375-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR