Imidazo[1,2-c]pyrimidine-2-carbonyl chloride ≥95%
Supplier: Matrix Scientific
083218-1G
102839-196EA
806.15
USD
102839-196
Imidazo[1,2-c]pyrimidine-2-carbonyl chloride ≥95%
Imidazo[1,2-c]pyrimidine-2-carbonyl chloride
Hazard: Irritant
Formula:
C₇H₄ClN₃O |
MDL Number:
MFCD08235244 CAS Number: 914637-85-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...