Imidazo[1,2-b]pyridazine-2-carbonyl chloride ≥95%

Supplier: Matrix Scientific

083216-1G
102839-192EA 806.15 USD
102839-192
Imidazo[1,2-b]pyridazine-2-carbonyl chloride ≥95%
Imidazo[1,2-b]pyridazine-2-carbonyl chloride

Hazard: Irritant

Formula: C₇H₄ClN₃O
MDL Number: MFCD08235242
CAS Number: 914637-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR