2-Methylindolizine-6-carbonyl chloride ≥97%

Supplier: Matrix Scientific

083215-1G
102839-190EA 750.77 USD
102839-190
2-Methylindolizine-6-carbonyl chloride ≥97%
2-Methylindolizine-6-carbonyl chloride

Hazard: Irritant

Formula: C₁₀H₈ClNO
MDL Number: MFCD08056328
CAS Number: 914637-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR