3,5-Dioxocyclohexanecarbonyl chloride ≥95%

Supplier: Matrix Scientific

083207-1G
102839-174EA 863.08 USD
102839-174
3,5-Dioxocyclohexanecarbonyl chloride ≥95%
3,5-Dioxocyclohexanecarbonyl chloride

Hazard: Irritant

Formula: C₇H₇ClO₃
MDL Number: MFCD21333126
CAS Number: 1355334-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR