4-Hydroxycyclohexanecarbonyl chloride ≥95%

Supplier: Matrix Scientific

083205-1G
102839-170EA 920 USD
102839-170
4-Hydroxycyclohexanecarbonyl chloride ≥95%
4-Hydroxycyclohexanecarbonyl chloride

Hazard: Irritant

Formula: C₇H₁₁ClO₂
MDL Number: MFCD18089216
CAS Number: 1174329-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR