3-Oxocyclobutanecarbonyl chloride ≥95%

Supplier: Matrix Scientific

083204-1G
102839-168EA 750.77 USD
102839-168
3-Oxocyclobutanecarbonyl chloride ≥95%
3-Oxocyclobutanecarbonyl chloride

Hazard: Flammable/Corrosive

Formula: C₅H₅ClO₂
MDL Number: MFCD12026359
CAS Number: 130233-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR