1-(3-Chlorobenzyl)-5-oxopyrrolidine-3-carbonyl chloride ≥97%
Supplier: Matrix Scientific
083195-1G
102839-150EA
949.23
USD
102839-150
1-(3-Chlorobenzyl)-5-oxopyrrolidine-3-carbonyl chloride ≥97%
1-(3-Chlorobenzyl)-5-oxopyrrolidine-3-carbonyl chloride
Hazard: Irritant
Formula:
C₁₂H₁₁Cl₂NO₂ |
MDL Number:
MFCD19706016 CAS Number: 1355334-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...