5-Methyl-1,2,3-oxadiazole-4-carbonyl chloride ≥95%

Supplier: Matrix Scientific

083194-1G
102839-148EA 707.69 USD
102839-148
5-Methyl-1,2,3-oxadiazole-4-carbonyl chloride ≥95%
5-Methyl-1,2,3-oxadiazole-4-carbonyl chloride

Hazard: Irritant

Formula: C₄H₃ClN₂O₂
MDL Number: MFCD08235239
CAS Number: 914637-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR