5-Chloro-1-methyl-1H-pyrazole-3-carbonyl chloride ≥95%
Supplier: Matrix Scientific
083175-1G
102839-104EA
863.08
USD
102839-104
5-Chloro-1-methyl-1H-pyrazole-3-carbonyl chloride ≥95%
5-Chloro-1-methyl-1H-pyrazole-3-carbonyl chloride
Hazard: Irritant
Formula:
C₅H₄Cl₂N₂O |
MDL Number:
MFCD11100096 CAS Number: 1017590-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...