2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetoyl chloride ≥97%

Supplier: Matrix Scientific

083172-1G
102839-096EA 949.23 USD
102839-096
2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetoyl chloride ≥97%
2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetoyl chloride

Hazard: Irritant

Formula: C₁₂H₁₀ClNO₂
MDL Number: MFCD03407529
CAS Number: 380648-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR