5-Thien-2-yl-1,3-oxazole-4-carbonyl chloride ≥95%

Supplier: Matrix Scientific

083171-1G
102839-094EA 707.69 USD
102839-094
5-Thien-2-yl-1,3-oxazole-4-carbonyl chloride ≥95%
5-Thien-2-yl-1,3-oxazole-4-carbonyl chloride

Hazard: Irritant

Formula: C₈H₄ClNO₂S
MDL Number: MFCD08056324
CAS Number: 914637-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR