5-(tert-Butyl)-1,3-oxazole-4-carbonyl chloride ≥95%

Supplier: Matrix Scientific

083168-1G
102839-088EA 806.15 USD
102839-088
5-(tert-Butyl)-1,3-oxazole-4-carbonyl chloride ≥95%
5-(tert-Butyl)-1,3-oxazole-4-carbonyl chloride

Hazard: Irritant

Formula: C₈H₁₀ClNO₂
MDL Number: MFCD08056321
CAS Number: 914637-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR