5-(Pyridin-3-yloxy)-2-furoyl chloride ≥95%
Supplier: Matrix Scientific
083155-1G
102839-056EA
666.15
USD
102839-056
5-(Pyridin-3-yloxy)-2-furoyl chloride ≥95%
5-(Pyridin-3-yloxy)-2-furoyl chloride
Hazard: Irritant
Formula:
C₁₀H₆ClNO₃ |
MDL Number:
MFCD03407386 CAS Number: 859850-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...