2-Chloro-6-(trifluoromethyl)nicotinoyl chloride ≥95%

Supplier: Matrix Scientific

083146-1G
102839-038EA 806.15 USD
102839-038
2-Chloro-6-(trifluoromethyl)nicotinoyl chloride ≥95%
2-Chloro-6-(trifluoromethyl)nicotinoyl chloride

Hazard: Irritant

Formula: C₇H₂Cl₂F₃NO
MDL Number: MFCD18089214
CAS Number: 1186306-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR