5-Chloro-2-methylnicotinoyl chloride ≥95%
Supplier: Matrix Scientific
083144-1G
102839-034EA
1315.38
USD
102839-034
5-Chloro-2-methylnicotinoyl chloride ≥95%
5-Chloro-2-methylnicotinoyl chloride
Hazard: Irritant
Formula:
C₇H₅Cl₂NO |
MDL Number:
MFCD16622246 CAS Number: 1207529-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...