3,5-Difluoro-4-(trifluoromethyl)benzoyl chloride ≥95%

Supplier: Matrix Scientific

083119-1G
102838-958EA 1118.46 USD
102838-958
3,5-Difluoro-4-(trifluoromethyl)benzoyl chloride ≥95%
3,5-Difluoro-4-(trifluoromethyl)benzoyl chloride

Hazard: Irritant

Formula: C₈H₂ClF₅O
MDL Number: MFCD01631627
CAS Number: 261945-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR