(2,6-Difluorophenyl)acetyl chloride ≥95%

Supplier: Matrix Scientific

083094-1G
102838-900EA 636.92 USD
102838-900
(2,6-Difluorophenyl)acetyl chloride ≥95%
(2,6-Difluorophenyl)acetyl chloride

Hazard: Irritant

Formula: C₈H₅ClF₂O
MW: 190.58 g/mol
MDL Number: MFCD03424718
CAS Number: 116622-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR