(5-Bromo-2-methylphenyl)acetyl chloride ≥95%

Supplier: Matrix Scientific

083085-1G
102838-880EA 1232.31 USD
102838-880
(5-Bromo-2-methylphenyl)acetyl chloride ≥95%
(5-Bromo-2-methylphenyl)acetyl chloride

Hazard: Irritant

Formula: C₉H₈BrClO
MDL Number: MFCD09038675
CAS Number: 1031387-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR