5-Bromopyridine-2-carbothioamide ≥97%

Supplier: Matrix Scientific

085072-1G
102844-172EA 920 USD
102844-172
5-Bromopyridine-2-carbothioamide ≥97%
5-Bromopyridine-2-carbothioamide

Hazard: Irritant

Formula: C₆H₅BrN₂S
MDL Number: MFCD12026374
CAS Number: 1215520-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR