6-Chloro-N-(4-ethylbenzyl)nicotinamide ≥97%

Supplier: Matrix Scientific

085070-1G
102844-168EA 553.85 USD
102844-168
6-Chloro-N-(4-ethylbenzyl)nicotinamide ≥97%
6-Chloro-N-(4-ethylbenzyl)nicotinamide

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂O
MDL Number: MFCD12026373
CAS Number: 1215716-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR