2-Chloro-N-(2-chloropyridin-3-yl)-4-fluorobenzamide ≥97%

Supplier: Matrix Scientific

085050-1G
102844-120EA 467.69 USD
102844-120
2-Chloro-N-(2-chloropyridin-3-yl)-4-fluorobenzamide ≥97%
2-Chloro-N-(2-chloropyridin-3-yl)-4-fluorobenzamide

Hazard: Irritant

Formula: C₁₂H₇Cl₂FN₂O
MDL Number: MFCD00206498
CAS Number: 648427-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR