4-(2-Methoxyphenyl)-3-nitrobut-3-en-2-one ≥97%
Supplier: Matrix Scientific
084946-1G
102843-796EA
1118.46
USD
102843-796
4-(2-Methoxyphenyl)-3-nitrobut-3-en-2-one ≥97%
4-(2-Methoxyphenyl)-3-nitrobut-3-en-2-one
Hazard: Irritant
Formula:
C₁₁H₁₁NO₄ |
MDL Number:
MFCD22566371 CAS Number: 1417574-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...