5-Chloro-2-(4-nitrophenoxy)benzonitrile ≥97%

Supplier: Matrix Scientific

084941-1G
102843-786EA 949.23 USD
102843-786
5-Chloro-2-(4-nitrophenoxy)benzonitrile ≥97%
5-Chloro-2-(4-nitrophenoxy)benzonitrile

Hazard: Irritant

Formula: C₁₃H₇ClN₂O₃
MDL Number: MFCD17982260
CAS Number: 1284752-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR