1-Nitro-4-[4-(trifluoromethoxy)phenoxy]benzene ≥97%

Supplier: Matrix Scientific

Synonyms: 1-nitro-4-(4-(trifluoromethoxy)phenoxy)benzene

084934-1G
102843-770EA 949.23 USD
102843-770
1-Nitro-4-[4-(trifluoromethoxy)phenoxy]benzene ≥97%
1-Nitro-4-[4-(trifluoromethoxy)phenoxy]benzene

Hazard: Irritant

Formula: C₁₃H₈F₃NO₄
MW: 299.21 g/mol
MDL Number: MFCD22126072
CAS Number: 74030-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR