3-(4-Nitrophenoxy)benzonitrile ≥97%

Supplier: Matrix Scientific

084929-1G
102843-760EA 949.23 USD
102843-760
3-(4-Nitrophenoxy)benzonitrile ≥97%
3-(4-Nitrophenoxy)benzonitrile

Hazard: Irritant

Formula: C₁₃H₈N₂O₃
MDL Number: MFCD19289635
CAS Number: 17076-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR