1-(2,4-Dinitrophenyl)-4-ethylpiperazine ≥97%

Supplier: Matrix Scientific

084921-1G
102843-742EA 806.15 USD
102843-742
1-(2,4-Dinitrophenyl)-4-ethylpiperazine ≥97%
1-(2,4-Dinitrophenyl)-4-ethylpiperazine

Hazard: Irritant

Formula: C₁₂H₁₆N₄O₄
MDL Number: MFCD01604345
CAS Number: 82448-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR