N'-Cyano-3-nitrobenzenecarboximidamide ≥97%
Supplier: Matrix Scientific
Synonyms:
N-Cyano-3-nitrobenzamidine
084920-1G
102843-740EA
863.08
USD
102843-740
N'-Cyano-3-nitrobenzenecarboximidamide ≥97%
N'-Cyano-3-nitrobenzenecarboximidamide
Hazard: Irritant
Formula:
C₈H₆N₄O₂ MW: 190.16 g/mol |
MDL Number:
MFCD22126071 CAS Number: 23275-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...