2,6-Dichloro-4-(1H-pyrrol-1-yl)phenol ≥97%

Supplier: Matrix Scientific

084911-1G
102843-714EA 750.77 USD
102843-714
2,6-Dichloro-4-(1H-pyrrol-1-yl)phenol ≥97%
2,6-Dichloro-4-(1H-pyrrol-1-yl)phenol

Hazard: Irritant

Formula: C₁₀H₇Cl₂NO
MDL Number: MFCD21333136
CAS Number: 855406-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR