3-Methyl-2-(1H-pyrrol-1-yl)phenol ≥97%
Supplier: Matrix Scientific
084902-1G
102843-690EA
806.15
USD
102843-690
3-Methyl-2-(1H-pyrrol-1-yl)phenol ≥97%
3-Methyl-2-(1H-pyrrol-1-yl)phenol
Hazard: Irritant
Formula:
C₁₁H₁₁NO |
MDL Number:
MFCD21333135 CAS Number: 1346497-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...