1-(2-tert-Butylphenyl)-1H-pyrrole ≥97%
Supplier: Matrix Scientific
084899-1G
102843-682EA
553.85
USD
102843-682
1-(2-tert-Butylphenyl)-1H-pyrrole ≥97%
1-(2-tert-Butylphenyl)-1H-pyrrole
Hazard: Irritant
Formula:
C₁₄H₁₇N |
MDL Number:
MFCD20141489 CAS Number: 1355334-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...