3-Amino-2-(1H-pyrrol-1-yl)phenol ≥97%

Supplier: Matrix Scientific

084896-1G
102843-672EA 949.23 USD
102843-672
3-Amino-2-(1H-pyrrol-1-yl)phenol ≥97%
3-Amino-2-(1H-pyrrol-1-yl)phenol

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MDL Number: MFCD21333133
CAS Number: 137352-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR