1-(3-Bromoprop-1-ynyl)-3,5-bis(trifluoromethyl)benzene ≥97%
Supplier: Matrix Scientific
084880-1G
102843-634EA
1118.46
USD
102843-634
1-(3-Bromoprop-1-ynyl)-3,5-bis(trifluoromethyl)benzene ≥97%
1-(3-Bromoprop-1-ynyl)-3,5-bis(trifluoromethyl)benzene
Hazard: Irritant
Formula:
C₁₁H₅BrF₆ |
MDL Number:
MFCD00119523 CAS Number: 81613-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...