tert-Butyl-4-(5-fluoro-2-nitrophenyl)piperazine-1-carboxylate ≥97%
Supplier: Matrix Scientific
084845-1G
102843-540EA
806.15
USD
102843-540
tert-Butyl-4-(5-fluoro-2-nitrophenyl)piperazine-1-carboxylate ≥97%
tert-Butyl-4-(5-fluoro-2-nitrophenyl)piperazine-1-carboxylate
Hazard: Irritant
Formula:
C₁₅H₂₀FN₃O₄ MW: 325.34 g/mol |
MDL Number:
MFCD18089261 CAS Number: 475279-74-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...