1-(5-Fluoro-2-nitrophenyl)piperidine ≥97%

Supplier: Matrix Scientific

084842-1G
102843-530EA 806.15 USD
102843-530
1-(5-Fluoro-2-nitrophenyl)piperidine ≥97%
1-(5-Fluoro-2-nitrophenyl)piperidine

Hazard: Irritant

Formula: C₁₁H₁₃FN₂O₂
MDL Number: MFCD18089260
CAS Number: 854044-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR