Cyclopropyl(2-fluorophenyl)methanol ≥97%

Supplier: Matrix Scientific

084830-1G
102843-502EA 863.08 USD
102843-502
Cyclopropyl(2-fluorophenyl)methanol ≥97%
Cyclopropyl(2-fluorophenyl)methanol

Hazard: Irritant

Formula: C₁₀H₁₁FO
MDL Number: MFCD16293485
CAS Number: 844470-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR