3-[Cyclopropyl(hydroxy)methyl]benzonitrile ≥97%

Supplier: Matrix Scientific

084829-1G
102843-500EA 636.92 USD
102843-500
3-[Cyclopropyl(hydroxy)methyl]benzonitrile ≥97%
3-[Cyclopropyl(hydroxy)methyl]benzonitrile

Hazard: Irritant

Formula: C₁₁H₁₁NO
MDL Number: MFCD18089257
CAS Number: 1002750-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR